missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-6-(trifluoromethyl)benzyl bromide, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442160010
Specifications
2-Fluoro-6-(trifluoromethyl)benzyl bromide | |
Glass Bottle | |
1g | |
94% min. (GC) | |
2-bromomethyl-1-fluoro-3-trifluoromethyl benzene, 2-bromomethyl-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethyl benzyl bromide, 2-fluoro-6-trifluoromethyl benzylbromide, 2-fluoro-6-trifluoromethylbenzyl bromide, 2-fluoro-6-trifluoromethylbenzylbromide, acmc-1cg83, alpha-bromo-2-fluoro-6-trifluoromethyl toluene, benzene, 2-bromomethyl-1-fluoro-3-trifluoromethyl, pubchem4932 | |
C1=CC(=C(C(=C1)F)CBr)C(F)(F)F | |
257.03 | |
257.03 |
107°C | |
42°C to 44°C | |
239087-08-2 | |
C8H5BrF4 | |
RINUERVPFANASB-UHFFFAOYSA-N | |
2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene | |
2737573 | |
95% |
Chemical Identifiers
239087-08-2 | |
257.03 | |
2-bromomethyl-1-fluoro-3-trifluoromethyl benzene, 2-bromomethyl-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethyl benzyl bromide, 2-fluoro-6-trifluoromethyl benzylbromide, 2-fluoro-6-trifluoromethylbenzyl bromide, 2-fluoro-6-trifluoromethylbenzylbromide, acmc-1cg83, alpha-bromo-2-fluoro-6-trifluoromethyl toluene, benzene, 2-bromomethyl-1-fluoro-3-trifluoromethyl, pubchem4932 | |
2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene |
C8H5BrF4 | |
RINUERVPFANASB-UHFFFAOYSA-N | |
2737573 | |
C1=CC(=C(C(=C1)F)CBr)C(F)(F)F |
Safety and Handling
GHS Signal Word: Danger