missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-6-(trifluoromethyl)benzoic acid, 98%, ACROS Organics™

Chemical Identifiers

CAS 32890-94-1
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.11
MDL Number MFCD00040981
InChI Key LNARMXLVVGHCRP-UHFFFAOYSA-N
Synonym 2-fluoro-6-trifluoromethyl benzoic acid, 2-fluoro-6-trifluoromethyl-benzoic acid, 6-fluoro-2-trifluoromethyl benzoic acid, 6-fluoro-2-trifluoromethylbenzoic acid, acmc-20a40d, alpha,alpha,alpha,6-tetrafluoro-o-toluic acid, benzoic acid, 2-fluoro-6-trifluoromethyl, otf-boa-6f, pubchem4759, rarechem al bo 0626
PubChem CID 688123
IUPAC Name 2-fluoro-6-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C(=C1)F)C(=O)O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC399920010 SDS View Documents Thermo Fisher Scientific
AC399920010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399920050 SDS View Documents Thermo Fisher Scientific
AC399920050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

32890-94-1
208.11
LNARMXLVVGHCRP-UHFFFAOYSA-N
688123
C1=CC(=C(C(=C1)F)C(=O)O)C(F)(F)F
C8H4F4O2
MFCD00040981
2-fluoro-6-trifluoromethyl benzoic acid, 2-fluoro-6-trifluoromethyl-benzoic acid, 6-fluoro-2-trifluoromethyl benzoic acid, 6-fluoro-2-trifluoromethylbenzoic acid, acmc-20a40d, alpha,alpha,alpha,6-tetrafluoro-o-toluic acid, benzoic acid, 2-fluoro-6-trifluoromethyl, otf-boa-6f, pubchem4759, rarechem al bo 0626
2-fluoro-6-(trifluoromethyl)benzoic acid

Specifications

Authentic
White to Yellow
1g
97.5
97.5% min. (HPLC)
FC6H3(CF3)CO2H
2-fluoro-6-trifluoromethyl benzoic acid, 2-fluoro-6-trifluoromethyl-benzoic acid, 6-fluoro-2-trifluoromethyl benzoic acid, 6-fluoro-2-trifluoromethylbenzoic acid, acmc-20a40d, alpha,alpha,alpha,6-tetrafluoro-o-toluic acid, benzoic acid, 2-fluoro-6-trifluoromethyl, otf-boa-6f, pubchem4759, rarechem al bo 0626
C1=CC(=C(C(=C1)F)C(=O)O)C(F)(F)F
208.11
208.11
98%
Glass bottle
85.0°C to 87.0°C
32890-94-1
100.0
C8H4F4O2
MFCD00040981
LNARMXLVVGHCRP-UHFFFAOYSA-N
2-fluoro-6-(trifluoromethyl)benzoic acid
688123
Crystalline Chunks
2-Fluoro-6-(trifluoromethyl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning