missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-(trifluoromethyl)benzeneboronic acid, 98%

$75.42 - $322.30

Chemical Identifiers

CAS 313545-34-5
Molecular Formula C7H5BF4O2
Molecular Weight (g/mol) 207.919
MDL Number MFCD07781239
InChI Key TXBRJTKFAYXKMV-UHFFFAOYSA-N
Synonym 2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenyl boronic acid, 2-fluoro-6-trifluoromethyl phenylboron&, 2-fluoro-6-trifluoromethylphenyl boronic acid
PubChem CID 16217168
IUPAC Name [2-fluoro-6-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=CC=C1F)C(F)(F)F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6414603 View Documents Alfa Aesar™
H6414603
1g Each for $75.42
 
AAH6414606 View Documents Alfa Aesar™
H6414606
5g Each for $322.30
 
Specifications

Chemical Identifiers

313545-34-5
207.919
TXBRJTKFAYXKMV-UHFFFAOYSA-N
16217168
B(C1=C(C=CC=C1F)C(F)(F)F)(O)O
C7H5BF4O2
MFCD07781239
2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenyl boronic acid, 2-fluoro-6-trifluoromethyl phenylboron&, 2-fluoro-6-trifluoromethylphenyl boronic acid
[2-fluoro-6-(trifluoromethyl)phenyl]boronic acid

Specifications

115°C to 119°C
313545-34-5
MFCD07781239
TXBRJTKFAYXKMV-UHFFFAOYSA-N
[2-fluoro-6-(trifluoromethyl)phenyl]boronic acid
16217168
Solid
2-Fluoro-6-(trifluoromethyl)benzeneboronic acid
1g
C7H5BF4O2
2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenyl boronic acid, 2-fluoro-6-trifluoromethyl phenylboron&, 2-fluoro-6-trifluoromethylphenyl boronic acid
B(C1=C(C=CC=C1F)C(F)(F)F)(O)O
207.919
207.92
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning