missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-(trifluoromethyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H6414606

 View more versions of this product

Catalog No. AAH6414606



Specifications

Specifications

2-Fluoro-6-(trifluoromethyl)benzeneboronic acid
5g
C7H5BF4O2
2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenyl boronic acid, 2-fluoro-6-trifluoromethyl phenylboron&, 2-fluoro-6-trifluoromethylphenyl boronic acid
B(C1=C(C=CC=C1F)C(F)(F)F)(O)O
207.919
207.92
98%
115°C to 119°C
313545-34-5
MFCD07781239
TXBRJTKFAYXKMV-UHFFFAOYSA-N
[2-fluoro-6-(trifluoromethyl)phenyl]boronic acid
16217168
Solid

Chemical Identifiers

313545-34-5
207.919
TXBRJTKFAYXKMV-UHFFFAOYSA-N
16217168
B(C1=C(C=CC=C1F)C(F)(F)F)(O)O
C7H5BF4O2
MFCD07781239
2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenyl boronic acid, 2-fluoro-6-trifluoromethyl phenylboron&, 2-fluoro-6-trifluoromethylphenyl boronic acid
[2-fluoro-6-(trifluoromethyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents