missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-6-(trifluoromethyl)benzaldehyde, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442270050
Specifications
2-Fluoro-6-(trifluoromethyl)benzaldehyde | |
156°C | |
60611-24-7 | |
C8H4F4O | |
FAKUGVHRTLCKHB-UHFFFAOYSA-N | |
2-fluoro-6-(trifluoromethyl)benzaldehyde | |
521933 | |
98% |
82°C | |
5g | |
97.5% min. (GC) | |
2-fluoro-6-trifluoromethyl benzaldehyde, 2-fluoro-6-trifluoromethyl-benzaldehyde, 3-fluoro-2-formylbenzotrifluoride, acmc-1b5by, alpha,alpha,alpha,6-tetrafluoro-o-tolualdehyde, attercop-chm at111495, benzaldehyde, 2-fluoro-6-trifluoromethyl, otf-bad-6f, pubchem4236 | |
C1=CC(=C(C(=C1)F)C=O)C(F)(F)F | |
192.11 | |
192.11 |
Chemical Identifiers
60611-24-7 | |
192.11 | |
2-fluoro-6-trifluoromethyl benzaldehyde, 2-fluoro-6-trifluoromethyl-benzaldehyde, 3-fluoro-2-formylbenzotrifluoride, acmc-1b5by, alpha,alpha,alpha,6-tetrafluoro-o-tolualdehyde, attercop-chm at111495, benzaldehyde, 2-fluoro-6-trifluoromethyl, otf-bad-6f, pubchem4236 | |
2-fluoro-6-(trifluoromethyl)benzaldehyde |
C8H4F4O | |
FAKUGVHRTLCKHB-UHFFFAOYSA-N | |
521933 | |
C1=CC(=C(C(=C1)F)C=O)C(F)(F)F |
Safety and Handling
GHS Signal Word: Warning