Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-(trifluoromethyl)benzaldehyde, 97%

$95.99 - $95.99

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzaldehyde
CAS 60611-24-7
Molecular Formula C8H4F4O
Formula Weight 192.11
MDL Number MFCD00061273
Beilstein 2263481
Synonym alpha,alpha,alpha,6-Tetrafluoro-o-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2065903

  SDS
alfa aesar™
B2065903
1g Each for $95.99
Description & Specifications

Specifications

Formula Weight 192.11
Assay 97%
Flash Point 81°C (177°F)
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzaldehyde
MDL Number MFCD00061273
Health Hazard 1 Warning
Quantity 1g
Molecular Formula C8H4F4O
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym alpha,alpha,alpha,6-Tetrafluoro-o-tolualdehyde
Beilstein 2263481
Boiling Point 155° to 157°C
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
CAS 60611-24-7
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.