Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-(trifluoromethyl)aniline, 98%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)aniline
CAS 144851-61-6
Molecular Formula C7H5F4N
Formula Weight 179.12
MDL Number MFCD00061242
UN Number UN1992
Synonym 2-Amino-3-fluorobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2068403

  SDS
alfa aesar™
B2068403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2068406

  SDS
alfa aesar™
B2068406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.12
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)aniline
Assay 98%
Molecular Formula C7H5F4N
MDL Number MFCD00061242
CAS 144851-61-6
UN Number UN1992
Synonym 2-Amino-3-fluorobenzotrifluoride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H226-H302-H332-H315-H319
Toxic in contact with skin.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
Flash Point 53°C (127°F)
Boiling Point 155°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H226-H302-H332-H315-H319
Toxic in contact with skin.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.