Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Fluoro-6'-(trifluoromethyl)acetophenone, 98%

$366.35 - $366.35

Chemical Identifiers

Chemical Name or Material 2'-Fluoro-6'-(trifluoromethyl)acetophenone
CAS 174013-29-7
Molecular Formula C9H6F4O
Formula Weight 206.14
MDL Number MFCD00061249
Beilstein 9193675
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2450106

 
alfa aesar™
B2450106
5g Each for $366.35
Description & Specifications

Specifications

Formula Weight 206.14
Chemical Name or Material 2'-Fluoro-6'-(trifluoromethyl)acetophenone
Assay 98%
Molecular Formula C9H6F4O
MDL Number MFCD00061249
CAS 174013-29-7
Beilstein 9193675
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 86°C (186°F)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.