Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-phenoxybenzonitrile, 95%

$48.34 - $166.24

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-phenoxybenzonitrile
CAS 175204-06-5
Molecular Formula C13H8FNO
Formula Weight 213.21
MDL Number MFCD00068201
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1456603

 
alfa aesar™
A1456603
1g Each for $48.34

AAA1456606

 
alfa aesar™
A1456606
5g Each for $166.24
Description & Specifications

Specifications

Formula Weight 213.21
Chemical Name or Material 2-Fluoro-6-phenoxybenzonitrile
Assay 95%
Molecular Formula C13H8FNO
MDL Number MFCD00068201
CAS 175204-06-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 90° to 93°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.