missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-phenoxybenzonitrile, 95%

Manufacturer:  Alfa Aesar™ A1456606

 View more versions of this product

Catalog No. AAA1456606



Specifications

Specifications

2-Fluoro-6-phenoxybenzonitrile
5g
C13H8FNO
2-fluoro-6-phenyloxybenzonitrile, 2-cyano-3-fluorodiphenyl ether, 6-fluoro-2-phenoxybenzenecarbonitrile, acmc-1bwys, maybridge1_005593, 2-fluoro-6-phenoxybenzonitrile, 2-fluoro-6-phenoxy benzonitrile, 2-fluoro-6-phenoxybenzonitrile, 2-fluoranyl-6-phenoxy-benzenecarbonitrile, 2-fluoro-6-phenoxybenzonitrile, tech
C1=CC=C(C=C1)OC2=C(C(=CC=C2)F)C#N
213.211
213.21
90°C to 93°C
175204-06-5
MFCD00068201
FBHXELGUZCMKQY-UHFFFAOYSA-N
2-fluoro-6-phenoxybenzonitrile
2737453
95%

Chemical Identifiers

175204-06-5
213.211
FBHXELGUZCMKQY-UHFFFAOYSA-N
2737453
C1=CC=C(C=C1)OC2=C(C(=CC=C2)F)C#N
C13H8FNO
MFCD00068201
2-fluoro-6-phenyloxybenzonitrile, 2-cyano-3-fluorodiphenyl ether, 6-fluoro-2-phenoxybenzenecarbonitrile, acmc-1bwys, maybridge1_005593, 2-fluoro-6-phenoxybenzonitrile, 2-fluoro-6-phenoxy benzonitrile, 2-fluoro-6-phenoxybenzonitrile, 2-fluoranyl-6-phenoxy-benzenecarbonitrile, 2-fluoro-6-phenoxybenzonitrile, tech
2-fluoro-6-phenoxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents