Learn More
Alfa Aesar™ 2-Fluoro-6-phenoxybenzonitrile, 95%
Manufacturer: Alfa Aesar™ A1456603
Specifications
2-Fluoro-6-phenoxybenzonitrile | |
1g | |
C13H8FNO | |
2-fluoro-6-phenyloxybenzonitrile, 2-cyano-3-fluorodiphenyl ether, 6-fluoro-2-phenoxybenzenecarbonitrile, acmc-1bwys, maybridge1_005593, 2-fluoro-6-phenoxybenzonitrile, 2-fluoro-6-phenoxy benzonitrile, 2-fluoro-6-phenoxybenzonitrile, 2-fluoranyl-6-phenoxy-benzenecarbonitrile, 2-fluoro-6-phenoxybenzonitrile, tech | |
C1=CC=C(C=C1)OC2=C(C(=CC=C2)F)C#N | |
213.211 | |
213.21 |
90°C to 93°C | |
175204-06-5 | |
MFCD00068201 | |
FBHXELGUZCMKQY-UHFFFAOYSA-N | |
2-fluoro-6-phenoxybenzonitrile | |
2737453 | |
95% |
Chemical Identifiers
175204-06-5 | |
213.211 | |
FBHXELGUZCMKQY-UHFFFAOYSA-N | |
2737453 | |
C1=CC=C(C=C1)OC2=C(C(=CC=C2)F)C#N |
C13H8FNO | |
MFCD00068201 | |
2-fluoro-6-phenyloxybenzonitrile, 2-cyano-3-fluorodiphenyl ether, 6-fluoro-2-phenoxybenzenecarbonitrile, acmc-1bwys, maybridge1_005593, 2-fluoro-6-phenoxybenzonitrile, 2-fluoro-6-phenoxy benzonitrile, 2-fluoro-6-phenoxybenzonitrile, 2-fluoranyl-6-phenoxy-benzenecarbonitrile, 2-fluoro-6-phenoxybenzonitrile, tech | |
2-fluoro-6-phenoxybenzonitrile |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning