Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-phenoxybenzaldehyde, 96%

$113.62 - $394.77

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-phenoxybenzaldehyde
CAS 902836-68-4
Molecular Formula C13H9FO2
Formula Weight 216.21
MDL Number MFCD03428585
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5084503

  SDS
alfa aesar™
H5084503
1g Each for $394.77

AAH50845MD

  SDS
alfa aesar™
H50845MD
250mg Each for $113.62
Description & Specifications

Specifications

Formula Weight 216.21
Chemical Name or Material 2-Fluoro-6-phenoxybenzaldehyde
Assay 96%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C13H9FO2
Health Hazard 1 Warning
Quantity 1g
MDL Number MFCD03428585
Melting Point 36° to 39°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 902836-68-4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.