Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-nitrotoluene, 98%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-nitrotoluene
CAS 769-10-8
Molecular Formula C7H6FNO2
Formula Weight 155.13
MDL Number MFCD00007197
UN Number UN2810
Beilstein 2361978
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2009306

  SDS
alfa aesar™
L2009306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2009314

  SDS
alfa aesar™
L2009314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2009322

  SDS
alfa aesar™
L2009322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.13
Chemical Name or Material 2-Fluoro-6-nitrotoluene
Assay 98%
MDL Number MFCD00007197
CAS 769-10-8
UN Number UN2810
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H6FNO2
Beilstein 2361978
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Flash Point 88°C (190°F)
Boiling Point 208° to 210°C
Melting Point 6° to 8°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.