Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-nitrobenzyl bromide, 98+%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-nitrobenzyl bromide
CAS 1958-93-6
Molecular Formula C7H5BrFNO2
Formula Weight 234.02
MDL Number MFCD00042918
UN Number UN3261
Beilstein 2503670
Synonym 2-Bromomethyl-1-fluoro-3-nitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1624006

  SDS
alfa aesar™
L1624006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1624003

  SDS
alfa aesar™
L1624003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 234.02
CAS 1958-93-6
MDL Number MFCD00042918
Health Hazard 1 Danger
Molecular Formula C7H5BrFNO2
UN Number UN3261
Chemical Name or Material 2-Fluoro-6-nitrobenzyl bromide
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 5g
Synonym 2-Bromomethyl-1-fluoro-3-nitrobenzene
Assay ≥98%
Beilstein 2503670
Melting Point 50° to 54°C
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Odor Pungent

Applications
Nitobenzyl ester based nonionic PAGs (NIPAG 2) NIPAG 2 was prepared from 2-fluoro-6-nitrobenzyl bromide with silver sulfonate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.