missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-methylbenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ H5522803

 View more versions of this product

Catalog No. AAH5522803



Specifications

Specifications

2-Fluoro-6-methylbenzaldehyde
1.525
1g
C8H7FO
2-fluoro-6-methyl-benzaldehyde
CC1=C(C(=CC=C1)F)C=O
138.141
138.14
97%
1.151 g/mL
Colorless to Yellow
117752-04-2
MFCD09258962
BGQBJWPZBBMKEI-UHFFFAOYSA-N
2-fluoro-6-methylbenzaldehyde
14170517
Liquid

Chemical Identifiers

117752-04-2
138.141
BGQBJWPZBBMKEI-UHFFFAOYSA-N
14170517
CC1=C(C(=CC=C1)F)C=O
C8H7FO
MFCD09258962
2-fluoro-6-methyl-benzaldehyde
2-fluoro-6-methylbenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents