missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-methylbenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ H55228MD

 View more versions of this product

Catalog No. AAH55228MD



Specifications

Specifications

2-Fluoro-6-methylbenzaldehyde
1.5250
250mg
C8H7FO
2-fluoro-6-methyl-benzaldehyde
CC1=C(C(=CC=C1)F)C=O
138.141
138.14
97%
1.151g/mL
Colorless to Yellow
117752-04-2
MFCD09258962
BGQBJWPZBBMKEI-UHFFFAOYSA-N
2-fluoro-6-methylbenzaldehyde
14170517
Liquid

Chemical Identifiers

117752-04-2
138.141
BGQBJWPZBBMKEI-UHFFFAOYSA-N
14170517
CC1=C(C(=CC=C1)F)C=O
C8H7FO
MFCD09258962
2-fluoro-6-methyl-benzaldehyde
2-fluoro-6-methylbenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents