missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-6-methoxybenzoic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436360010
Specifications
2-Fluoro-6-methoxybenzoic acid | |
Glass bottle | |
85°C to 87°C | |
137654-21-8 | |
100.0 | |
C8H7FO3 | |
2-fluoro-6-methoxy-benzoic acid, 2-fluoro-6-methoxybenzoicacid, 6-fluoro-2-anisoic acid, acmc-209cem, benzoic acid, 2-fluoro-6-methoxy, intermediates-zcf02063, ksc494o0r, methyl-6-fluorosalicylic acid, pubchem4967, rarechem al bo 0442 | |
COC1=C(C(=CC=C1)F)C(=O)O | |
170.14 | |
170.14 | |
97% |
Authentic | |
White | |
1g | |
96.0 | |
96% min. (HPLC) | |
MFCD00671764 | |
XLNQGZLCGVLNMF-UHFFFAOYSA-N | |
2-fluoro-6-methoxybenzoic acid | |
2737360 | |
Crystalline Powder |
Chemical Identifiers
137654-21-8 | |
170.14 | |
XLNQGZLCGVLNMF-UHFFFAOYSA-N | |
2737360 | |
COC1=C(C(=CC=C1)F)C(=O)O |
C8H7FO3 | |
MFCD00671764 | |
2-fluoro-6-methoxy-benzoic acid, 2-fluoro-6-methoxybenzoicacid, 6-fluoro-2-anisoic acid, acmc-209cem, benzoic acid, 2-fluoro-6-methoxy, intermediates-zcf02063, ksc494o0r, methyl-6-fluorosalicylic acid, pubchem4967, rarechem al bo 0442 | |
2-fluoro-6-methoxybenzoic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning