missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-6-methoxybenzoic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC436360050

 View more versions of this product

Catalog No. AC436360050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Fluoro-6-methoxybenzoic acid
Glass bottle
85°C to 87°C
137654-21-8
100.0
C8H7FO3
2-fluoro-6-methoxy-benzoic acid, 2-fluoro-6-methoxybenzoicacid, 6-fluoro-2-anisoic acid, acmc-209cem, benzoic acid, 2-fluoro-6-methoxy, intermediates-zcf02063, ksc494o0r, methyl-6-fluorosalicylic acid, pubchem4967, rarechem al bo 0442
COC1=C(C(=CC=C1)F)C(=O)O
170.14
170.14
97%
Authentic
White
5g
96.0
96% min. (HPLC)
MFCD00671764
XLNQGZLCGVLNMF-UHFFFAOYSA-N
2-fluoro-6-methoxybenzoic acid
2737360
Crystalline Powder

Chemical Identifiers

137654-21-8
170.14
XLNQGZLCGVLNMF-UHFFFAOYSA-N
2737360
COC1=C(C(=CC=C1)F)C(=O)O
C8H7FO3
MFCD00671764
2-fluoro-6-methoxy-benzoic acid, 2-fluoro-6-methoxybenzoicacid, 6-fluoro-2-anisoic acid, acmc-209cem, benzoic acid, 2-fluoro-6-methoxy, intermediates-zcf02063, ksc494o0r, methyl-6-fluorosalicylic acid, pubchem4967, rarechem al bo 0442
2-fluoro-6-methoxybenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents