Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-methoxybenzoic acid, 97%

$49.73 - $148.72

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-methoxybenzoic acid
CAS 137654-21-8
Molecular Formula C8H7FO3
Formula Weight 170.14
MDL Number MFCD00671764
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2606003

  SDS
alfa aesar™
H2606003
1g Each for $49.73

AAH2606006

  SDS
alfa aesar™
H2606006
5g Each for $148.72
Description & Specifications

Specifications

Formula Weight 170.14
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Melting Point 89° to 93°C
CAS 137654-21-8
Assay 97%
Molecular Formula C8H7FO3
MDL Number MFCD00671764
Chemical Name or Material 2-Fluoro-6-methoxybenzoic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.