missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 2-Fluoro-6-methoxybenzeneboronic acid pinacol ester, 96%
Chemical Identifiers
CAS | 1599432-41-3 |
---|---|
Molecular Formula | C13H18BFO3 |
Molecular Weight (g/mol) | 252.092 |
InChI Key | CSXSULPDIWBLHT-UHFFFAOYSA-N |
Synonym | 2-2-fluoro-6-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-6-methoxybenzeneboronic acid pinacol ester, 2-fluoro-6-methoxybenzeneboronicacidpinacolester, 2-fluoro-6-methoxyphenylboronic acid 2,3-dimethyl-2,3-butanediyl, 2-fluoro-6-methoxyphenylboronic acid pinacol ester, 2-fluoro-6-methoxyphenylboronicacidpinacolester |
PubChem CID | 72221147 |
IUPAC Name | 2-(2-fluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC=C2F)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH62812MD
|
Alfa Aesar™
H62812MD |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AAH6281203
|
Alfa Aesar™
H6281203 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1599432-41-3 | |
252.092 | |
2-2-fluoro-6-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-6-methoxybenzeneboronic acid pinacol ester, 2-fluoro-6-methoxybenzeneboronicacidpinacolester, 2-fluoro-6-methoxyphenylboronic acid 2,3-dimethyl-2,3-butanediyl, 2-fluoro-6-methoxyphenylboronic acid pinacol ester, 2-fluoro-6-methoxyphenylboronicacidpinacolester | |
2-(2-fluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
C13H18BFO3 | |
CSXSULPDIWBLHT-UHFFFAOYSA-N | |
72221147 | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC=C2F)OC |
Specifications
250mg | |
C13H18BFO3 | |
CSXSULPDIWBLHT-UHFFFAOYSA-N | |
2-(2-fluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
72221147 | |
Solid | |
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester |
1599432-41-3 | |
2-2-fluoro-6-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-6-methoxybenzeneboronic acid pinacol ester, 2-fluoro-6-methoxybenzeneboronicacidpinacolester, 2-fluoro-6-methoxyphenylboronic acid 2,3-dimethyl-2,3-butanediyl, 2-fluoro-6-methoxyphenylboronic acid pinacol ester, 2-fluoro-6-methoxyphenylboronicacidpinacolester | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC=C2F)OC | |
252.092 | |
252.09 | |
96% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures