missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-6-methoxybenzaldehyde, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC435460010
Specifications
2-Fluoro-6-methoxybenzaldehyde | |
Authentic | |
Tan to White | |
1g | |
97.5 | |
97.5% min. (GC) | |
2-fluoro-6-mehoxybenzaldehyde, 2-fluoro-6-methoxy-benzaldehyde, 2-fluoro-6-methoxybenzaldehyde, 2-methoxy-6-fluorobenzaldehyde, 3-fluoro-2-formylanisole, 6-fluoro-o-anisaldehyde, acmc-209yud, benzaldehyde, 2-fluoro-6-methoxy, intermediates-zcf02117, pubchem8486 | |
COC1=C(C(=CC=C1)F)C=O | |
154.14 | |
154.14 | |
98% |
110°C | |
Glass Bottle | |
57.0°C to 60.0°C | |
146137-74-8 | |
100.0 | |
C8H7FO2 | |
UIOAYOIJMYMOEU-UHFFFAOYSA-N | |
2-fluoro-6-methoxybenzaldehyde | |
2737357 | |
Crystalline Powder |
Chemical Identifiers
146137-74-8 | |
154.14 | |
2-fluoro-6-mehoxybenzaldehyde, 2-fluoro-6-methoxy-benzaldehyde, 2-fluoro-6-methoxybenzaldehyde, 2-methoxy-6-fluorobenzaldehyde, 3-fluoro-2-formylanisole, 6-fluoro-o-anisaldehyde, acmc-209yud, benzaldehyde, 2-fluoro-6-methoxy, intermediates-zcf02117, pubchem8486 | |
2-fluoro-6-methoxybenzaldehyde |
C8H7FO2 | |
UIOAYOIJMYMOEU-UHFFFAOYSA-N | |
2737357 | |
COC1=C(C(=CC=C1)F)C=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning