Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-methoxybenzaldehyde, 98%

$39.26 - $39.26

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-methoxybenzaldehyde
CAS 146137-74-8
Molecular Formula C8H7FO2
Formula Weight 154.14
MDL Number MFCD01090998
Synonym 6-Fluoro-o-anisaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26086MD

 
alfa aesar™
H26086MD
250mg Each for $39.26
Description & Specifications

Specifications

Formula Weight 154.14
Chemical Name or Material 2-Fluoro-6-methoxybenzaldehyde
Assay 98%
Molecular Formula C8H7FO2
MDL Number MFCD01090998
CAS 146137-74-8
Synonym 6-Fluoro-o-anisaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Melting Point 58° to 60°C
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.