missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-iodotoluene, 97%

$37.92 - $450.01

Chemical Identifiers

CAS 443-85-6
Molecular Formula C7H6FI
Molecular Weight (g/mol) 236.028
MDL Number MFCD00142922
InChI Key MSPXWJMFEVAKHQ-UHFFFAOYSA-N
Synonym 1-fluoranyl-3-iodanyl-2-methyl-benzene
PubChem CID 2774523
IUPAC Name 1-fluoro-3-iodo-2-methylbenzene
SMILES CC1=C(C=CC=C1I)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55149MD View Documents Alfa Aesar™
H55149MD
250mg Each for $37.92
 
AAH5514903 View Documents Alfa Aesar™
H5514903
1g Each for $110.57
 
AAH5514906 View Documents Alfa Aesar™
H5514906
5g Each for $450.01
 
Specifications

Chemical Identifiers

443-85-6
236.028
MSPXWJMFEVAKHQ-UHFFFAOYSA-N
2774523
CC1=C(C=CC=C1I)F
C7H6FI
MFCD00142922
1-fluoranyl-3-iodanyl-2-methyl-benzene
1-fluoro-3-iodo-2-methylbenzene

Specifications

1.808
1.583
117°C (9mmHg)
443-85-6
MFCD00142922
MSPXWJMFEVAKHQ-UHFFFAOYSA-N
1-fluoro-3-iodo-2-methylbenzene
2774523
97%
84°C (183°F)
Light sensitive
250mg
C7H6FI
1-fluoranyl-3-iodanyl-2-methyl-benzene
CC1=C(C=CC=C1I)F
236.028
236.03
2-Fluoro-6-iodotoluene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H227-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures