Please login to your online account to display your discounted pricing

2-Fluoro-6-iodobenzoyl chloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-iodobenzoyl chloride
CAS 111771-12-1
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H3ClFIO
Linear Formula FC6H3(I)COCl
Formula Weight 284.45
MDL Number MFCD03094410
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC426130050

  SDS
thermo fisher scientific
AC426130050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 284.45
Quantity 5g
Density 1.956g/mL
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Molecular Formula C7H3ClFIO
Assay Percent Range 97%
CAS 111771-12-1
Chemical Name or Material 2-Fluoro-6-iodobenzoyl chloride
Linear Formula FC6H3(I)COCl
MDL Number MFCD03094410
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.