missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-fluoro-6-iodobenzonitrile 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC380740010

 View more versions of this product

Catalog No. AC380740010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Fluoro-6-iodobenzonitrile
49° to 53°C
79544-29-9
100.0
Glass bottle
≥97.5%
C7H3FIN
247.01
White
1g
97.5
Authentic
Crystals or Crystalline Powder
98%
IC6H3(F)CN
MFCD00015478

Chemical Identifiers

79544-29-9
100.0
C7H3FIN
247.01
2-Fluoro-6-iodobenzonitrile
97.5
98%
IC6H3(F)CN
MFCD00015478
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning