missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-fluoro-6-iodobenzonitrile 98%, ACROS Organics™

Chemical Identifiers

CAS 79544-29-9
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H3FIN
Linear Formula IC6H3(F)CN
Formula Weight 247.01
MDL Number MFCD00015478
Chemical Name or Material 2-Fluoro-6-iodobenzonitrile
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC380740010 SDS Thermo Fisher Scientific
AC380740010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

79544-29-9
100.0
C7H3FIN
247.01
2-Fluoro-6-iodobenzonitrile
97.5
98%
IC6H3(F)CN
MFCD00015478
≥97.5%

Specifications

Crystals or Crystalline Powder
1g
49° to 53°C
IC6H3(F)CN
Glass bottle
C7H3FIN
97.5
2-Fluoro-6-iodobenzonitrile
247.01
White
100.0
Authentic
MFCD00015478
98%
79544-29-9
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning