Please login to your online account to display your discounted pricing

2-Fluoro-6-iodobenzoic acid, 98%, ACROS Organics™

$71.90 - $71.90

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-iodobenzoic acid
Name Note 98%
CAS 111771-08-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4FIO2
Linear Formula FC6H3(I)CO2H
Formula Weight 266
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC311940050

  SDS
thermo fisher scientific
AC311940050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC311940010

  SDS
thermo fisher scientific
311940010
Glass bottle 1g Each for $71.90
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 266
Quantity 5g
Color Yellow to light brown
TSCA TSCA
Melting Point 121° to 126°C
CAS Max % 100.0
Chemical Name or Material 2-Fluoro-6-iodobenzoic acid
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H4FIO2
Linear Formula FC6H3(I)CO2H
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
MDL Number MFCD00042289
CAS 111771-08-5
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.