Please login to your online account to display your discounted pricing

2-Fluoro-6-iodobenzoic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  311940010

 View more versions of this product

Catalog No. AC311940010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 266
Physical Form Powder
Quantity 1g
Color Yellow to light brown
TSCA TSCA
Melting Point 121° to 126°C
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Infrared Spectrum Authentic
Molecular Formula C7H4FIO2
Assay Percent Range 98%
Chemical Name or Material 2-Fluoro-6-iodobenzoic acid
CAS 111771-08-5
Name Note 98%
Linear Formula FC6H3(I)CO2H
MDL Number MFCD00042289

Specifications

Chemical Name or Material 2-Fluoro-6-iodobenzoic acid
Name Note 98%
CAS 111771-08-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4FIO2
Linear Formula FC6H3(I)CO2H
Formula Weight 266
MDL Number MFCD00042289
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents