Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-hydroxypyridine, 97%

$106.66 - $297.08

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-hydroxypyridine
CAS 50543-23-2
Molecular Formula C5H4FNO
Formula Weight 113.09
MDL Number MFCD03092938
Synonym 6-Fluoro-2-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3168603

 
alfa aesar™
H3168603
1g Each for $297.08

AAH31686MD

 
alfa aesar™
H31686MD
250mg Each for $106.66
Description & Specifications

Specifications

Formula Weight 113.09
Chemical Name or Material 2-Fluoro-6-hydroxypyridine
Assay 97%
Molecular Formula C5H4FNO
MDL Number MFCD03092938
CAS 50543-23-2
Synonym 6-Fluoro-2-pyridinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 125° to 129°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.