Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-6-hydroxybenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-hydroxybenzeneboronic acid pinacol ester
Formula Weight 238.06
Synonym 2-Fluoro-6-hydroxyphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6237403

 
alfa aesar™
H6237403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62374MD

 
alfa aesar™
H62374MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 238.06
Physical Form Solid
Synonym 2-Fluoro-6-hydroxyphenylboronic acid pinacol ester
Chemical Name or Material 2-Fluoro-6-hydroxybenzeneboronic acid pinacol ester
Assay 96%
Quantity 1g