missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-(trifluoromethyl)pyridine 98.0+%, TCI America™

$22.80 - $65.36

Chemical Identifiers

CAS 69045-82-5
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD07437945
InChI Key UUODQIKUTGWMPT-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-5-trifluoromethyl-pyridine, acmc-209o5o, alpha,alpha,alpha,6-tetrafluoro-3-picoline, ksc628s4n, pubchem9252, pyridine, 2-fluoro-5-trifluoromethyl
PubChem CID 5463754
IUPAC Name 2-fluoro-5-(trifluoromethyl)pyridine
SMILES C1=CC(=NC=C1C(F)(F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F08461G
View Documents Promotion Details
Tci America
F08461G
1g
Each for $22.80
 
F08465G
View Documents Promotion Details
Tci America
F08465G
5g
Each for $65.36
 
Description

Description

Specifications

Chemical Identifiers

69045-82-5
165.091
UUODQIKUTGWMPT-UHFFFAOYSA-N
5463754
C1=CC(=NC=C1C(F)(F)F)F
C6H3F4N
MFCD07437945
2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-5-trifluoromethyl-pyridine, acmc-209o5o, alpha,alpha,alpha,6-tetrafluoro-3-picoline, ksc628s4n, pubchem9252, pyridine, 2-fluoro-5-trifluoromethyl
2-fluoro-5-(trifluoromethyl)pyridine

Specifications

120°C
1g
C6H3F4N
1993
UUODQIKUTGWMPT-UHFFFAOYSA-N
2-fluoro-5-(trifluoromethyl)pyridine
5463754
Clear Liquid at 20°C
2-Fluoro-5-(trifluoromethyl)pyridine
Yellow
69045-82-5
MFCD07437945
2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-5-trifluoromethyl-pyridine, acmc-209o5o, alpha,alpha,alpha,6-tetrafluoro-3-picoline, ksc628s4n, pubchem9252, pyridine, 2-fluoro-5-trifluoromethyl
C1=CC(=NC=C1C(F)(F)F)F
165.091
165.09
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

SDS
Product Certifications