Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Fluoro-5'-(trifluoromethyl)propiophenone, 97%

$76.50 - $301.70

Chemical Identifiers

Chemical Name or Material 2'-Fluoro-5'-(trifluoromethyl)propiophenone
CAS 207974-18-3
Molecular Formula C10H8F4O
Formula Weight 220.17
MDL Number MFCD00061260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2066003

 
alfa aesar™
B2066003
1g Each for $76.50

AAB2066006

 
alfa aesar™
B2066006
5g Each for $301.70
Description & Specifications

Specifications

Formula Weight 220.17
Chemical Name or Material 2'-Fluoro-5'-(trifluoromethyl)propiophenone
Assay 97%
Molecular Formula C10H8F4O
MDL Number MFCD00061260
CAS 207974-18-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.