missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%, ACROS Organics™

$64.75 - $64.75

Chemical Identifiers

CAS 69922-27-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H3F4NO
Linear Formula FC6H3(CF3)nCO
Formula Weight 205.11
MDL Number MFCD00673069
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC319340025 SDS Thermo Fisher Scientific
AC319340025
Glass bottle 2.5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC319345000 SDS Thermo Fisher Scientific
319345000
Glass Bottle 500mg Each for $64.75
Specifications Description & Specifications

Chemical Identifiers

69922-27-6
100.0
C8H3F4NO
205.11
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%
96.0
97%
FC6H3(CF3)nCO
MFCD00673069
≥96.0%

Specifications

Liquid
2.5g
100.0
Authentic
Glass bottle
97%
MFCD00673069
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%
205.11
Clear colorless
96.0
1.4540 to 1.4560 (20°C, 589nm)
C8H3F4NO
FC6H3(CF3)nCO
69922-27-6
≥96.0%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA