missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 319345000

 View more versions of this product

Catalog No. AC319345000


 

Specifications Description & Specifications

Specifications

≥96.0%
205.11
Clear colorless
100.0
1.4540 to 1.4560 (20°C, 589nm)
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate
97%
C8H3F4NO
69922-27-6
Liquid
500mg
Glass Bottle
96.0
Authentic
MFCD00673069
FC6H3(CF3)nCO
97%

Chemical Identifiers

69922-27-6
100.0
C8H3F4NO
205.11
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%
96.0
97%
FC6H3(CF3)nCO
MFCD00673069
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA