Please login to your online account to display your discounted pricing

2-Fluoro-5-(trifluoromethyl)phenol, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC397840050

 View more versions of this product

Catalog No. AC397840050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 180.1
Quantity 5g
Color Clear colorless to orange
Density 1.4360g/mL
CAS Max % 100.0
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)phenol
Refractive Index 1.4380 to 1.4400 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C7H4F4O
Linear Formula FC6H3(OH)CF3
CAS 141483-15-0
Infrared Spectrum Authentic
MDL Number MFCD00061286
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)phenol
CAS 141483-15-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4F4O
Linear Formula FC6H3(OH)CF3
Formula Weight 180.1
MDL Number MFCD00061286
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents