Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(trifluoromethyl)phenol, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)phenol
CAS 141483-15-0
Molecular Formula C7H4F4O
Formula Weight 180.1
MDL Number MFCD00061286
UN Number UN2920
Beilstein 8392138
Synonym alpha,alpha,alpha,6-Tetrafluoro-m-cresol; 4-Fluoro-3-hydroxybenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2000403

 
alfa aesar™
B2000403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2000406

 
alfa aesar™
B2000406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.1
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)phenol
Assay 97%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318-H226-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.
Flash Point 57°C (134°F)
Boiling Point 146°C
Molecular Formula C7H4F4O
MDL Number MFCD00061286
CAS 141483-15-0
UN Number UN2920
Beilstein 8392138
Synonym alpha,alpha,alpha,6-Tetrafluoro-m-cresol; 4-Fluoro-3-hydroxybenzotrifluoride
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H226-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.