Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(trifluoromethyl)benzophenone, 96%

$433.95 - $433.95

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzophenone
CAS 199292-40-5
Molecular Formula C14H8F4O
Formula Weight 268.21
MDL Number MFCD00061254
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1993706

  SDS
alfa aesar™
A1993706
5g Each for $433.95
Description & Specifications

Specifications

Formula Weight 268.21
MDL Number MFCD00061254
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C14H8F4O
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Health Hazard 1 Warning
Assay 96%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzophenone
CAS 199292-40-5
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.