Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(trifluoromethyl)benzoic acid, 98+%

$50.92 - $162.73

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzoic acid
CAS 115029-23-7
Molecular Formula C8H4F4O2
Formula Weight 208.11
MDL Number MFCD00061292
Beilstein 5813056
Synonym alpha,alpha,alpha,6-Tetrafluoro-m-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2029303

  SDS
alfa aesar™
B2029303
1g Each for $50.92

AAB2029306

  SDS
alfa aesar™
B2029306
5g Each for $162.73
Description & Specifications

Specifications

Formula Weight 208.11
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzoic acid
CAS 115029-23-7
Beilstein 5813056
Synonym alpha,alpha,alpha,6-Tetrafluoro-m-toluic acid
Assay ≥98%
MDL Number MFCD00061292
Melting Point 101° to 102°C
Quantity 1g
Molecular Formula C8H4F4O2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.