Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(trifluoromethyl)benzaldehyde, 97%

$42.01 - $454.08

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzaldehyde
CAS 146137-78-2
Molecular Formula C8H4F4O
Formula Weight 192.11
MDL Number MFCD00061311
Beilstein 5810686
Synonym alpha,alpha,alpha,6-Tetrafluoro-m-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1997303

 
alfa aesar™
A1997303
1g Each for $42.01

AAA1997306

 
alfa aesar™
A1997306
5g Each for $105.58

AAA1997314

 
alfa aesar™
A1997314
25g Each for $454.08
Description & Specifications

Specifications

Formula Weight 192.11
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzaldehyde
Assay 97%
Molecular Formula C8H4F4O
MDL Number MFCD00061311
CAS 146137-78-2
Beilstein 5810686
Synonym alpha,alpha,alpha,6-Tetrafluoro-m-tolualdehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 76°C (168°F)
Boiling Point 176°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.