Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Fluoro-5'-(trifluoromethyl)acetophenone, 97+%

$97.49 - $355.59

Chemical Identifiers

Chemical Name or Material 2'-Fluoro-5'-(trifluoromethyl)acetophenone
CAS 202664-53-7
Molecular Formula C9H6F4O
Formula Weight 206.14
MDL Number MFCD00061248
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2077703

  SDS
alfa aesar™
B2077703
1g Each for $97.49

AAB2077706

  SDS
alfa aesar™
B2077706
5g Each for $355.59
Description & Specifications

Specifications

Formula Weight 206.14
Molecular Formula C9H6F4O
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥97%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
MDL Number MFCD00061248
CAS 202664-53-7
Chemical Name or Material 2'-Fluoro-5'-(trifluoromethyl)acetophenone
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.