Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(trifluoromethoxy)phenylacetic acid, 97%

$93.42 - $290.81

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)phenylacetic acid
CAS 886497-98-9
Molecular Formula C9H6F4O3
Formula Weight 238.14
MDL Number MFCD04115892
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2671403

  SDS
alfa aesar™
H2671403
1g Each for $290.81

AAH26714MD

  SDS
alfa aesar™
H26714MD
250mg Each for $93.42
Description & Specifications

Specifications

Formula Weight 238.14
Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)phenylacetic acid
Assay 97%
Molecular Formula C9H6F4O3
MDL Number MFCD04115892
CAS 886497-98-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 54° to 56°C
Quantity 1g

Applications
As intermediates. Commonly used laboratory reagent. Used in the field of nanoparticles.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.