Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(trifluoromethoxy)benzoic acid, 97%

$105.03 - $575.58

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)benzoic acid
CAS 886497-85-4
Molecular Formula C8H4F4O3
Formula Weight 224.11
MDL Number MFCD04115887
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2633903

  SDS
alfa aesar™
H2633903
1g Each for $105.03

AAH2633909

  SDS
alfa aesar™
H2633909
10g Each for $575.58
Description & Specifications

Specifications

Formula Weight 224.11
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 78° to 81°C
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD04115887
Quantity 1g
Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)benzoic acid
Molecular Formula C8H4F4O3
Health Hazard 1 Warning
CAS 886497-85-4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.