Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(trifluoromethoxy)benzaldehyde, 97%

$129.92 - $1,168.31

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)benzaldehyde
CAS 886497-81-0
Molecular Formula C8H4F4O2
Formula Weight 208.11
MDL Number MFCD04115886
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2665503

  SDS
alfa aesar™
H2665503
1g Each for $129.92

AAH2665506

  SDS
alfa aesar™
H2665506
5g Each for $383.28

AAH2665514

  SDS
alfa aesar™
H2665514
25g Each for $1,168.31
Description & Specifications

Specifications

Formula Weight 208.11
Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)benzaldehyde
Assay 97%
Molecular Formula C8H4F4O2
MDL Number MFCD04115886
CAS 886497-81-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
2-Fluoro-5-(trifluoromethoxy)benzaldehyde is used as a precursor for organic synthesis. It is also used as a pharmaceutical intermediate.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents, strong reducing agents, strong acids, ammonia and bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.