Learn More
2-Fluoro-5-(trifluoromethoxy)aniline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H3210303
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIn discovery of novel benzimidazoles as potent inhibitors of TIE-2 and VEGFR-2 tyrosine kinase receptors 2-Fluoro-5-(trifluoromethoxy)aniline is used as coupling agent. 2-Fluoro-5-(trifluoromethyl)aniline may be used in chemical synthesis studies.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
2-Fluoro-5-(trifluoromethoxy)aniline | |
C7H5F4NO | |
MFCD09832254 | |
UN2810 | |
Insoluble in water. | |
NC1=C(F)C=CC(OC(F)(F)F)=C1 | |
195.12 | |
195.12 |
116369-23-4 | |
1.4485 | |
1 g | |
2-fluoro-5-trifluoromethoxy aniline, benzenamine,2-fluoro-5-trifluoromethoxy, benzenamine, 2-fluoro-5-trifluoromethoxy, acmc-20adyv, pubchem12670, synquest 4656-3-37, 2-fluoro-3-trifluoromethoxy aniline, 2-fluoro-5-trifluoromethoxy phenylamine, 2-fluoro-5-trifluoromethoxy aniline,jrd | |
HQUWXEXDIIWBBY-UHFFFAOYSA-N | |
2-fluoro-5-(trifluoromethoxy)aniline | |
14439308 | |
97% |
Chemical Identifiers
116369-23-4 | |
195.12 | |
HQUWXEXDIIWBBY-UHFFFAOYSA-N | |
2-fluoro-5-(trifluoromethoxy)aniline |
C7H5F4NO | |
MFCD09832254 | |
14439308 | |
NC1=C(F)C=CC(OC(F)(F)F)=C1 |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H302+H332-H311-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures