missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-nitrotoluene, 98+%

Manufacturer:  Alfa Aesar™ A1464909

 View more versions of this product

Catalog No. AAA1464909



Specifications

Specifications

2-Fluoro-5-nitrotoluene
98°C to 101°C (13mmHg)
10g
C7H6FNO2
UN2811
2-fluoro-5-nitrotoluene, benzene, 1-fluoro-2-methyl-4-nitro, 4-fluoro-3-methylnitrobenzene, toluene, 2-fluoro-5-nitro, 1-fluoro-2-methyl-4-nitro-benzene, 5-nitro-2-fluorotoluene, 3-methyl-4-fluoronitrobenzene, pubchem1593, 2-fluoro-5-nitro-toluene, 5-nitro-2-fluoro toluene
CC1=C(C=CC(=C1)[N+](=O)[O-])F
155.128
155.13
105°C (221°F)
37°C to 40°C
455-88-9
MFCD00007284
1940341
XUCYJGMIICONES-UHFFFAOYSA-N
1-fluoro-2-methyl-4-nitrobenzene
68001
≥98%

Chemical Identifiers

455-88-9
155.128
XUCYJGMIICONES-UHFFFAOYSA-N
68001
CC1=C(C=CC(=C1)[N+](=O)[O-])F
C7H6FNO2
MFCD00007284
2-fluoro-5-nitrotoluene, benzene, 1-fluoro-2-methyl-4-nitro, 4-fluoro-3-methylnitrobenzene, toluene, 2-fluoro-5-nitro, 1-fluoro-2-methyl-4-nitro-benzene, 5-nitro-2-fluorotoluene, 3-methyl-4-fluoronitrobenzene, pubchem1593, 2-fluoro-5-nitro-toluene, 5-nitro-2-fluoro toluene
1-fluoro-2-methyl-4-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

Documents