Please login to your online account to display your discounted pricing

2-Fluoro-5-nitrotoluene, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-nitrotoluene
CAS 455-88-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H6FNO2
Linear Formula CH3C6H3(NO2)F
Formula Weight 155.13
MDL Number MFCD00007284
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC119510100

  SDS
thermo fisher scientific
119510100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystals or Crystalline Chunks
Formula Weight 155.13
Quantity 10g
Color Yellow
Melting Point 36° to 41°C
Beilstein 05, II, 251
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Fluoro-5-nitrotoluene
Molecular Formula C7H6FNO2
MDL Number MFCD00007284
CAS 455-88-9
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula CH3C6H3(NO2)F
Packaging Glass bottle
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.