missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-nitrobenzonitrile, 98+%

Manufacturer:  Alfa Aesar™ A1241706

 View more versions of this product

Catalog No. AAA1241706


 

Specifications

Specifications

2-Fluoro-5-nitrobenzonitrile
5g
≥98%
MFCD00042299
2048720
C1=CC(=C(C=C1[N+](=O)[O-])C#N)F
166.111
166.11
70°C to 72°C
17417-09-3
C7H3FN2O2
UN3439
YLACBMHBZVYOAP-UHFFFAOYSA-N
2-fluoro-5-nitrobenzonitrile
519417

Chemical Identifiers

17417-09-3
166.111
YLACBMHBZVYOAP-UHFFFAOYSA-N
C7H3FN2O2
MFCD00042299
519417
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents