Please login to your online account to display your discounted pricing

2-Fluoro-5-nitrobenzonitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-nitrobenzonitrile
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H3FN2O2
Formula Weight 166.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00351DA

 
fisher scientific company
TL00351DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.11
Quantity 1g
Assay 97%
Chemical Name or Material 2-Fluoro-5-nitrobenzonitrile
Assay Percent Range 97%
Molecular Formula C7H3FN2O2
CAS 17417-09-3