Please login to your online account to display your discounted pricing

2-Fluoro-5-nitrobenzonitrile, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  TL00351DA

 View more versions of this product

Catalog No. TL00351DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 166.11
Quantity 1g
Assay 97%
Chemical Name or Material 2-Fluoro-5-nitrobenzonitrile
Assay Percent Range 97%
CAS 17417-09-3
Molecular Formula C7H3FN2O2

Specifications

Chemical Name or Material 2-Fluoro-5-nitrobenzonitrile
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H3FN2O2
Formula Weight 166.11