missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-nitrobenzoic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC398170050

 View more versions of this product

Catalog No. AC398170050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Fluoro-5-nitrobenzoic acid
138° to 142°C
Authentic
97.5
Glass bottle
≥97.5%
C7H4FNO4
185.11
White to pale yellow
5g
7304-32-7
100.0
Crystalline Powder
98%
NO2C6H3(F)COOH
MFCD00134238

Chemical Identifiers

7304-32-7
100.0
C7H4FNO4
185.11
2-Fluoro-5-nitrobenzoic acid
97.5
98%
NO2C6H3(F)COOH
MFCD00134238
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.